Connection system GL 45, DURAN®
Screw cap, GL 45, PP, with two ports GL 14
554-3000
Connection system with PP screw cap GL 45 with two or three PTFE ports. Flexible modular system. Typical applications: safe transfer of liquid media within a closed and sterile system (evaporation is reduced).
- Four tubing sizes (1,6, 3,0, 3,2 or 6,0 mm) can be used by selecting the correct insert
- Sterile pressure equalisation is possible through use of membrane filter
- Unused ports can be provided with a blind cap
- Descrizione:Screw cap, GL 45, PP, with two ports GL 14
Accessories
- Descrizione
- Descrizione
- Descrizione
- Descrizione
- Descrizione
- Descrizione
- 554-3009Kit per equalizzazione della pressione per tappo a vite a 2 e 3 porte, compl. di membrana filtrante da 0,2 μm1 * 1 ST - SFr. 37.90
- Descrizione