pH Combination electrodes
Sensolyt® SEA-HP pH single rod measuring chain**, Sensolyt® SEA-HP, to be installed into the SensoLyt® Fitting, 4 - 12, Increased pressure/temperature conditions
Codice catalogo WTWO109118
Nome del fornitore: WTW ONLINE
Vai a
Product Details & Documents
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
- Ideal for measurements in water and drinking water
- Also for high pressure and temperature conditions
- Easy to maintain by easy measuring chain replacement
Specifiche
- Per:Increased pressure/temperature conditions
- Tipo:Sensolyt® SEA-HP, to be installed into the SensoLyt® Fitting
- Descrizione:Sensolyt® SEA-HP pH single rod measuring chain**
- Seq:36
- pH:4 - 12