Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Bromocresolo

Glass bottle for solids, 25 g
Codice catalogo A17090.14
Nome del fornitore:   Thermo Fisher Scientific
Bromocresolo
Codice catalogo A17090.14
Nome del fornitore:  Thermo Fisher Scientific
Numero CAS:  76-60-8
Bromocresolo
Bromocresolo

Glass bottle for solids, 25 g

Numero CAS:  76-60-8
Price
SFr. 247.00
Log in per disponibilità
1 * 25 g
Product Details & Documents

CAS No.: 76-60-8 Molecular Formula: C21H14Br4O5S Formula Weight: 698.02 Melting Point: ca 225° dec. Beilstein Reference: 372527 Merck Reference: 14,1386 MDL No.: MFCD00005874

Sinonimi:  3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • Numero CAS:76-60-8
  • Formula:C₂₁H₁₄Br₄O₅S
  • Numero MDL:MFCD00005874
  • Molecular Weight:698.02 g/mol
  • IndexMerck:14,1386
  • Punto di fusione:∼ 225 °C (dec.)
  • temperatura di conservazione:Ambiente
  • EINECS:200-972-8
Specifiche

Specifiche

  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    25 g

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.