Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Timolo ≥98%

500 g
Catalog # A14563.36
Corrosive
Irritant
Environment
Timolo ≥98%
Catalog # A14563.36
CAS Number:  89-83-8
Timolo ≥98%
Timolo ≥98%

500 g

CAS Number:  89-83-8
Price
SFr. 171.00
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98+% CAS No.: 89-83-8 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 102°(215°F) pH: 5-7 Melting Point: 49-52° Boiling Point: 231-233° Beilstein Reference: 1907135 Merck Reference: 14,9399 UN No.: 2430 MDL No.: MFCD00002309

Synonyms:  4-Isopropyl-3-hydroxytoluene, 2-Isopropyl-5-methylphenol, 6-Isopropyl-m-cresol
  • CAS Number:89-83-8
  • Formula:2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
  • MDL Number:MFCD00002309
  • Molecular Weight:150.22 g/mol
  • Merck Index:14,9399
  • Boiling Pt:231…233 °C
  • Melting Pt:49…52 °C
  • Density:0,970
  • Flash Pt:102 °C (215 °F)
  • Storage Temperature:Ambiente
  • EINECS:201-944-8
  • UN:2430
  • ADR:8,III
Specifications

Specifications

  • Conf:
    500 g

Specifications