Orders LinkContactUsLinkComponent
Fumaroyl chloride 95%
Fumaroyl chloride 95%
Glass bottle for solids, 25 g
  A14540.14
Fumaroyl chloride 95%
  A14540.14
 :  627-63-4
Fumaroyl chloride 95%
Fumaroyl chloride 95%
Glass bottle for solids, 25 g
 :  627-63-4
SFr. 96.40
1 * 25 g
Purity: 95% CAS No.: 627-63-4 Molecular Formula: C4H2Cl2O2 Formula Weight: 152.97 Flash Point: 73°(163°F) Refractive Index: 1.5000 Odour: Acrid Melting Point: -2° Boiling Point: 159-161° Beilstein Reference: 1721342 UN No.: 1780 MDL No.: MFCD00000733
 :  Fumaryl chloride
  •  :627-63-4
  •  :C₄H₂Cl₂O₂
  •  :MFCD00000733
  •  :152.96 g/mol
  •  :159…161 °C
  •  :–2 °C
  •  :1,413
  •  :73 °C (163 °F)
  •  :Ambiente
  •  :211-005-4
  •  :1780
  •  :8,II
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    25 g