Fumaroyl chloride 95%
Glass bottle for solids, 25 g
Purity: 95% CAS No.: 627-63-4 Molecular Formula: C4H2Cl2O2 Formula Weight: 152.97 Flash Point: 73°(163°F) Refractive Index: 1.5000 Odour: Acrid Melting Point: -2° Boiling Point: 159-161° Beilstein Reference: 1721342 UN No.: 1780 MDL No.: MFCD00000733
: Fumaryl chloride
- :627-63-4
- :C₄H₂Cl₂O₂
- :MFCD00000733
- :152.96 g/mol
- :159…161 °C
- :–2 °C
- :1,413
- :73 °C (163 °F)
- :Ambiente
- :211-005-4
- :1780
- :8,II
Documents
- Tipo di conf.:Glass bottle for solids
- Conf:25 g