1,3-Dihydroxyacetone dimer ≥97%
Glass bottle for solids, 50 g
Nome del fornitore: Thermo Fisher Scientific
1,3-Dihydroxyacetone dimer ≥97%
Codice catalogo A14189.18
Nome del fornitore: Thermo Fisher Scientific
Numero CAS: 62147-49-3
Vai a
Product Details & Documents
Purity: 97% CAS No.: 62147-49-3 Molecular Formula: C6H12O6 Formula Weight: 180.16 Storage Temperature: 2°C to 8°C Melting Point: ca 75-82° Beilstein Reference: 112910 Merck Reference: 14,3182 MDL No.: MFCD00051019
Sinonimi: 2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol
- Numero CAS:62147-49-3
- Formula:C₆H₁₂O₆
- Numero MDL:MFCD00051019
- Molecular Weight:180.16 g/mol
- IndexMerck:14,3182
- Punto di fusione:∼ 75…82 °C
- EINECS:612-947-2
Documents
Specifiche
Specifiche
- Tipo di conf.:Glass bottle for solids
- Conf:50 g
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



