Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

3-Nitrobenzaldehyde ≥99%

Bottiglia di vetro, 250 g
Catalog # A13594.30
Irritant
3-Nitrobenzaldehyde ≥99%
Catalog # A13594.30
CAS Number:  99-61-6
3-Nitrobenzaldehyde ≥99%
3-Nitrobenzaldehyde ≥99%

Bottiglia di vetro, 250 g

CAS Number:  99-61-6
Price
SFr. 108.00
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 99-61-6 Molecular Formula: C7H5NO3 Formula Weight: 151.12 Melting Point: 57-59° Boiling Point: 164°/23mm Merck Reference: 14,6587 MDL No.: MFCD00007249

  • CAS Number:99-61-6
  • Formula:C₇H₅NO₃
  • MDL Number:MFCD00007249
  • Molecular Weight:151.12 g/mol
  • Merck Index:14,6587
  • Boiling Pt:164 °C (23 mmHg)
  • Melting Pt:57…59 °C
  • Density:1,481 g/cm³ (0 °C)
  • Storage Temperature:Ambiente
  • EINECS:202-772-6
Specifications

Specifications

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    250 g

Specifications