Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Fmoc-Pro-OH ≥98%

Glass bottle for solids, 25 g
Catalog # B21081.14
Fmoc-Pro-OH ≥98%
Catalog # B21081.14
CAS Number:  71989-31-6
Fmoc-Pro-OH ≥98%
Fmoc-Pro-OH ≥98%

Glass bottle for solids, 25 g

CAS Number:  71989-31-6
Price
378.00 SFr.
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 71989-31-6 Molecular Formula: C20H19NO4 Formula Weight: 337.38 Storage Temperature: 2°C to 8°C Melting Point: 113-117° Beilstein Reference: 3596735 Optical Rotation: -40° (c=1 in ethyl acetate) MDL No.: MFCD00037122

Synonyms:  N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline, N-Fmoc-L-proline
  • CAS Number:71989-31-6
  • Formula:C₂₀H₁₉NO₄
  • MDL Number:MFCD00037122
  • Molecular Weight:337.38 g/mol
  • Melting Pt:113…117 °C
  • EINECS:276-259-0
Specifications

Specifications

  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    25 g

Specifications