Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Solfanilammide ≥98%

Solfanilammide ≥98%

Glass bottle for solids, 500 g
Catalog # A13001.36
Irritant
Solfanilammide ≥98%
Catalog # A13001.36
CAS Number:  63-74-1
Solfanilammide ≥98%
Solfanilammide ≥98%

Glass bottle for solids, 500 g

CAS Number:  63-74-1
Price
158.00 SFr.
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98% CAS No.: 63-74-1 Molecular Formula: C6H8N2O2S Formula Weight: 172.21 Melting Point: 163-168° Beilstein Reference: 511852 Merck Reference: 14,8925 MDL No.: MFCD00007939

Synonyms:  4-Aminobenzenesulphonamide
  • CAS Number:63-74-1
  • Formula:H₂NC₆H₄SO₂NH₂
  • MDL Number:MFCD00007939
  • Molecular Weight:172.21 g/mol
  • Merck Index:14,8925
  • Boiling Pt:401 °C (1013 hPa)
  • Melting Pt:163…168 °C
  • Density:1,08
  • Storage Temperature:Ambiente
  • EINECS:200-563-4
Specifications

Specifications

  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.