Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Etilendiammina 99%

Etilendiammina 99%

bottiglia in vetro, 2.500 mL
Catalog # A12132.0F
Health
Corrosive
Flammable
Irritant
Etilendiammina 99%
Catalog # A12132.0F
CAS Number:  107-15-3
Etilendiammina 99%
Etilendiammina 99%

bottiglia in vetro, 2.500 mL

CAS Number:  107-15-3
Price
SFr. 158.00
Log in for availability
1 * 2.500 mL
Product Details & Documents

Purity: 99% CAS No.: 107-15-3 Molecular Formula: C2H8N2 Formula Weight: 60.10 Flash Point: 38°(100°F) Physical Form: Liquid Density: 0.899 Refractive Index: 1.4565 Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Odour: Ammonia-like Melting Point: 8-11° Boiling Point: 117-118° Solubility: Soluble in water, alcohol. Slightly soluble in ether Beilstein Reference: 605263 Merck Reference: 14,3795 UN No.: 1604 MDL No.: MFCD00008204

Strongly basic amine useful as a building block in chemical synthesis

Synonyms:  1,2-Ethanediamine, 1,2-Diaminoethane
  • CAS Number:107-15-3
  • Formula:C₂H₈N₂
  • MDL Number:MFCD00008204
  • Molecular Weight:60.1 g/mol
  • Merck Index:14,3795
  • Boiling Pt:117…118 °C
  • Melting Pt:8…11 °C
  • Density:0,899
  • Flash Pt:38 °C (100 °F)
  • Storage Temperature:Ambiente
  • EINECS:203-468-6
  • UN:1604
  • ADR:8,II
Specifications

Specifications

  • Tipo di conf.:
    bottiglia in vetro
  • Conf:
    2.500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.