Orders LinkContactUsLinkComponent
D-(-)-Penicillamine 98%
Bottiglia di vetro, 100 g
  A11446.22
D-(-)-Penicillamine 98%
  A11446.22
 :  52-67-5
D-(-)-Penicillamine 98%
D-(-)-Penicillamine 98%
Bottiglia di vetro, 100 g
 :  52-67-5
SFr. 1’790.00
1 * 100 g
Purity: 99% CAS No.: 52-67-5 Molecular Formula: C5H11NO2S Formula Weight: 149.21 Storage Temperature: 2°C to 8°C Odour: Unpleasant pH: 4.5-5.5 Melting Point: 202-204° dec. Beilstein Reference: 1722375 Merck Reference: 14,7088 Optical Rotation: -63° (c=1 in 1N NaOH) MDL No.: MFCD00064302
 :  H-D-Pen-OH, (S)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine
  •  :52-67-5
  •  :C₅H₁₁NO₂S
  •  :MFCD00064302
  •  :149.2 g/mol
  •  :14,7088
  •  :202…204 °C (dec.)
  •  :200-148-8
  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    100 g