Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
D-(-)-Penicillamine 98%

D-(-)-Penicillamine 98%

Bottiglia di vetro, 25 g
N° de catalogue A11446.14
Irritant
Fournisseur:   Thermo Fisher Scientific
D-(-)-Penicillamine 98%
N° de catalogue A11446.14
Numéro CAS:  52-67-5
D-(-)-Penicillamine 98%
D-(-)-Penicillamine 98%

Bottiglia di vetro, 25 g

Numéro CAS:  52-67-5
Prix
SFr. 536.00
Connexion Pour la disponibilité
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 52-67-5 Molecular Formula: C5H11NO2S Formula Weight: 149.21 Storage Temperature: 2°C to 8°C Odour: Unpleasant pH: 4.5-5.5 Melting Point: 202-204° dec. Beilstein Reference: 1722375 Merck Reference: 14,7088 Optical Rotation: -63° (c=1 in 1N NaOH) MDL No.: MFCD00064302

Synonymes:  H-D-Pen-OH, (S)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine
  • Numéro CAS:52-67-5
  • Formule:C₅H₁₁NO₂S
  • Numéro MDL:MFCD00064302
  • Poids moléculaire:149.2 g/mol
  • Index Merck:14,7088
  • Point de fusion:202…204 °C (dec.)
  • EINECS:200-148-8
Spécifications

Spécifications

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    25 g

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.