D-(-)-Penicillamine 98%
Bottiglia di vetro, 5 g
Purity: 99% CAS No.: 52-67-5 Molecular Formula: C5H11NO2S Formula Weight: 149.21 Storage Temperature: 2°C to 8°C Odour: Unpleasant pH: 4.5-5.5 Melting Point: 202-204° dec. Beilstein Reference: 1722375 Merck Reference: 14,7088 Optical Rotation: -63° (c=1 in 1N NaOH) MDL No.: MFCD00064302
: H-D-Pen-OH, (S)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine
- :52-67-5
- :C₅H₁₁NO₂S
- :MFCD00064302
- :149.2 g/mol
- :14,7088
- :202…204 °C (dec.)
- :200-148-8
Documents
- Tipo di conf.:Bottiglia di vetro
- Conf:5 g