Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
2-Acetylthiazole ≥99%

2-Acetylthiazole ≥99%

Glass bottle for solids, 1 g
Catalog # A11266.03
Irritant
2-Acetylthiazole ≥99%
Catalog # A11266.03
CAS Number:  24295-03-2
2-Acetylthiazole ≥99%
2-Acetylthiazole ≥99%

Glass bottle for solids, 1 g

CAS Number:  24295-03-2
Price
124.00 SFr.
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 99% CAS No.: 24295-03-2 Molecular Formula: C5H5NOS Formula Weight: 127.17 Flash Point: 78°(172°F) Refractive Index: 1.5480 Odour: Characteristic Boiling Point: 89-91°/12mm Beilstein Reference: 109803 MDL No.: MFCD00005324

Synonyms:  1-(Thiazol-2-yl)-1-ethanone
  • CAS Number:24295-03-2
  • Formula:C₅H₅NOS
  • MDL Number:MFCD00005324
  • Molecular Weight:127.17 g/mol
  • Boiling Pt:89…91 °C (12 mmHg)
  • Melting Pt:65,5 °C
  • Density:1,227
  • Flash Pt:78 °C (172 °F)
  •  :Densità: 1,22 g/cm³ (20 °C)
  • Storage Temperature:Ambiente
  • EINECS:246-134-5
Specifications

Specifications

  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.