Orders LinkContactUsLinkComponent
Veratraldehyde ≥99%
Bottiglia di vetro, 500 g
  A11110.36
Veratraldehyde ≥99%
  A11110.36
 :  120-14-9
Veratraldehyde ≥99%
Veratraldehyde ≥99%
Bottiglia di vetro, 500 g
 :  120-14-9
SFr. 166.00
1 * 500 g
Purity: 99% CAS No.: 120-14-9 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 112°(233°F) Melting Point: 42-46° Boiling Point: 281° Beilstein Reference: 473899 Merck Reference: 14,9951 MDL No.: MFCD00003363
 :  3,4-Dimethoxybenzaldehyde
  •  :120-14-9
  •  :C₉H₁₀O₃
  •  :MFCD00003363
  •  :166.18 g/mol
  •  :14,9951
  •  :281 °C
  •  :42…46 °C
  •  :112 °C (233 °F)
  •  :Ambiente
  •  :204-373-2
  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    500 g