Veratraldehyde ≥99%
Bottiglia di vetro, 100 g
Purity: 99% CAS No.: 120-14-9 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 112°(233°F) Melting Point: 42-46° Boiling Point: 281° Beilstein Reference: 473899 Merck Reference: 14,9951 MDL No.: MFCD00003363
: 3,4-Dimethoxybenzaldehyde
- :120-14-9
- :C₉H₁₀O₃
- :MFCD00003363
- :166.18 g/mol
- :14,9951
- :281 °C
- :42…46 °C
- :112 °C (233 °F)
- :Ambiente
- :204-373-2
Documents
- Tipo di conf.:Bottiglia di vetro
- Conf:100 g