Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
EDTA 99%

EDTA 99%

Bottiglia di vetro, 1.000 g
Catalog # A10713.0B
Irritant
EDTA 99%
Catalog # A10713.0B
CAS Number:  60-00-4
EDTA 99%
EDTA 99%

Bottiglia di vetro, 1.000 g

CAS Number:  60-00-4
Price
SFr. 132.00
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 99% CAS No.: 60-00-4 Molecular Formula: C10H16N2O8 Formula Weight: 292.24 Melting Point: ca 245° dec. Beilstein Reference: 1716295 Merck Reference: 14,3517 MDL No.: MFCD00003541

Synonyms:  Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
  • CAS Number:60-00-4
  • Formula:C₁₀H₁₆N₂O₈
  • MDL Number:MFCD00003541
  • Molecular Weight:292.25 g/mol
  • Merck Index:14,3517
  • Boiling Pt:614 °C (1013 hPa)
  • Melting Pt:∼ 245 °C (dec.)
  • Density:0,86 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambiente
  • EINECS:200-449-4
  • REACH:01-2119486399-18
Specifications

Specifications

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    1.000 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.