Orders LinkContactUsLinkComponent
Ammonio persolfato (APS) ≥98%
Glass bottle for solids, 500 g
  A10533.36
Ammonio persolfato (APS) ≥98%
  A10533.36
 :  7727-54-0
Ammonio persolfato (APS) ≥98%
Ammonio persolfato (APS) ≥98%
Glass bottle for solids, 500 g
 :  7727-54-0
SFr. 49.90
1 * 500 g
Purity: 98% CAS No.: 7727-54-0 Molecular Formula: H8N2O8S2 Formula Weight: 228.19 Flash Point: None Refractive Index: 1.50 Odour: Odourless Melting Point: 120° dec. Merck Reference: 14,541 UN No.: 1444 MDL No.: MFCD00003390
 :  Peroxidisulphuric acid diammonium salt, Ammonium persulphate
  •  :7727-54-0
  •  :(NH₄)₂S₂O₈
  •  :MFCD00003390
  •  :228.2 g/mol
  •  :14,541
  •  :120 °C (dec.)
  •  :1,982
  •  :Ambiente
  •  :231-786-5
  •  :1444
  •  :5.1,III
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    500 g