Ammonio persolfato (APS) ≥98%
Glass bottle for solids, 5.000 g
Purity: 98% CAS No.: 7727-54-0 Molecular Formula: H8N2O8S2 Formula Weight: 228.19 Flash Point: None Refractive Index: 1.50 Odour: Odourless Melting Point: 120° dec. Merck Reference: 14,541 UN No.: 1444 MDL No.: MFCD00003390
: Peroxidisulphuric acid diammonium salt, Ammonium persulphate
- :7727-54-0
- :(NH₄)₂S₂O₈
- :MFCD00003390
- :228.2 g/mol
- :14,541
- :120 °C (dec.)
- :1,982
- :Ambiente
- :231-786-5
- :1444
- :5.1,III
- Tipo di conf.:Glass bottle for solids
- Conf:5.000 g