Orders LinkContactUsLinkComponent
Bifenile ≥99%
Glass bottle for solids, 5.000 g
  A10265.0I
Bifenile ≥99%
  A10265.0I
 :  92-52-4
Bifenile ≥99%
Bifenile ≥99%
Glass bottle for solids, 5.000 g
 :  92-52-4
SFr. 178.00
1 * 5.000 g
Purity: 99% CAS No.: 92-52-4 Molecular Formula: C12H10 Formula Weight: 154.21 Flash Point: 113°(235°F) Odour: Pleasant Boiling Point: 254-255° Solubility: Soluble in alcohol and ether Beilstein Reference: 1634058 Merck Reference: 14,3314 UN No.: 3077 MDL No.: MFCD00003054
 :  Diphenyl, Phenylbenzene, PCB No. 0, 0-PCB
  •  :92-52-4
  •  :C₁₂H₁₀
  •  :MFCD00003054
  •  :154.21 g/mol
  •  :14,3314
  •  :254…255 °C
  •  :69…72 °C
  •  :0,990
  •  :113 °C (235 °F)
  •  :Ambiente
  •  :202-163-5
  •  :3077
  •  :9,III
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    5.000 g