Bifenile ≥99%
Glass bottle for solids, 5.000 g
Purity: 99% CAS No.: 92-52-4 Molecular Formula: C12H10 Formula Weight: 154.21 Flash Point: 113°(235°F) Odour: Pleasant Boiling Point: 254-255° Solubility: Soluble in alcohol and ether Beilstein Reference: 1634058 Merck Reference: 14,3314 UN No.: 3077 MDL No.: MFCD00003054
: Diphenyl, Phenylbenzene, PCB No. 0, 0-PCB
- :92-52-4
- :C₁₂H₁₀
- :MFCD00003054
- :154.21 g/mol
- :14,3314
- :254…255 °C
- :69…72 °C
- :0,990
- :113 °C (235 °F)
- :Ambiente
- :202-163-5
- :3077
- :9,III
- Tipo di conf.:Glass bottle for solids
- Conf:5.000 g