Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Bifenile ≥99%

Bifenile ≥99%

Glass bottle for solids, 1.000 g
N° de catalogue A10265.0B
Irritant
Environnement
Fournisseur:   Thermo Fisher Scientific
Bifenile ≥99%
N° de catalogue A10265.0B
Numéro CAS:  92-52-4
Bifenile ≥99%
Bifenile ≥99%

Glass bottle for solids, 1.000 g

Numéro CAS:  92-52-4
Prix
SFr. 69.10
Connexion Pour la disponibilité
1 * 1.000 g
Product Details & Documents

Purity: 99% CAS No.: 92-52-4 Molecular Formula: C12H10 Formula Weight: 154.21 Flash Point: 113°(235°F) Odour: Pleasant Boiling Point: 254-255° Solubility: Soluble in alcohol and ether Beilstein Reference: 1634058 Merck Reference: 14,3314 UN No.: 3077 MDL No.: MFCD00003054

Synonymes:  Diphenyl, Phenylbenzene, PCB No. 0, 0-PCB
  • Numéro CAS:92-52-4
  • Formule:C₁₂H₁₀
  • Numéro MDL:MFCD00003054
  • Poids moléculaire:154.21 g/mol
  • Index Merck:14,3314
  • Point d'ébullition:254…255 °C
  • Point de fusion:69…72 °C
  • Densité:0,990
  • Point d'éclair:113 °C (235 °F)
  • Température de stockage:Ambiente
  • EINECS:202-163-5
  • UN:3077
  • ADR:9,III
Spécifications

Spécifications

  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    1.000 g

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.