Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

3',5'-Dihydroxyacetophenone 96%

Glass bottle for solids, 10 g
Catalog # B25567.09
Irritant
3',5'-Dihydroxyacetophenone 96%
Catalog # B25567.09
CAS Number:  51863-60-6
3',5'-Dihydroxyacetophenone 96%
3',5'-Dihydroxyacetophenone 96%

Glass bottle for solids, 10 g

CAS Number:  51863-60-6
Price
SFr. 215.00
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 96% CAS No.: 51863-60-6 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 142-149° Beilstein Reference: 1936502

Synonyms:  1-(3,5-Dihydroxyphenyl)ethan-1-one, 1-(3,5-Dihydroxyphenyl)ethanone
  • CAS Number:51863-60-6
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00002290
  • Molecular Weight:152.15 g/mol
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:142…149 °C
  • Storage Temperature:Ambiente
  • EINECS:257-480-1
Specifications

Specifications

  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.