Heating tapes, HBS
Heating tape HBS, 7 m length, 1550 W
Catalog # HORS020308
Supplier: HORST
Jump to:
Product Details & Documents
The heating tapes are smooth, flexible and have a multiple glass-yarn insulation as well as an additional protection braid around the heating conductors.
Specifications
- Descrizione:Heating tape HBS, 7 m length, 1550 W
- Seq:08