Disposable cannulas
Disposable cannulas, G14, verde, 70 mm, +, 2×70 mm
Catalog # EHRS15886470
Supplier: EHRHARDT SOEHNE
Jump to:
Product Details & Documents
Disposable cannulas.
- Pyrogen-free, not toxic
- With lancet cut
- EO-sterilisation
Documents
Certificates
Search by Lot NumberSpecifications
- Ø×L:2×70 mm
- Gauge:G14
- Sterili:+
- Colore:verde
- Descrizione:Disposable cannulas
- Lunghezza:70 mm
- Seq:11