Orders LinkContactUsLinkComponent
Heating tapes, HBS
Heating tapes, HBS
Heating tape HBS, 2 m length, 500 W
Codice catalogo HORS020304
Nome del fornitore:   HORST
Heating tapes, HBS
Codice catalogo HORS020304
Nome del fornitore:  HORST
Heating tapes, HBS
Heating tape HBS, 2 m length, 500 W
Price
167.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
The heating tapes are smooth, flexible and have a multiple glass-yarn insulation as well as an additional protection braid around the heating conductors.
Specifiche
  • Descrizione:
    Heating tape HBS, 2 m length, 500 W
  • Seq:
    03