Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Glass beads

Glass beads

Glass beads, solid, 4 mm
Codice catalogo LENZ05124004
Nome del fornitore:   Lenz Laborglas
Glass beads
Codice catalogo LENZ05124004
Nome del fornitore:  Lenz Laborglas
Glass beads
Glass beads

Glass beads, solid, 4 mm

Price
67.10 SFr.
Log in per disponibilità
1 * 1 kg
Product Details & Documents

Solid soda-lime glass.

Specifiche
  • Ø:
    4 mm
  • Descrizione:
    Glass beads, solid
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.