Bottles with pump vaporiser
30 ml, GL 18 thread, ambra
Codice catalogo 215-6272
Nome del fornitore: KOEHLER TECHNISCHE PRODUKTEN
Bottles with pump vaporiser
Codice catalogo 215-6272
Nome del fornitore: KOEHLER TECHNISCHE PRODUKTEN
Vai a
Product Details & Documents
Amber soda glass bottles with pump vaporiser and suitable for chromatography.
- Pump volume 0,12 ml per stroke
- Good chemical resistance
- Amber coloured for light sensitive substances
Specifiche
- Tipo:GL 18 thread
- Capacità:30 ml
- Colore:ambra
- Seq:03