Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Storage Boxes, PP, with Lid | Avantor

Storage Boxes, PP, with Lid

with transparent hinged lid, 10×10 compartments, blu, 155×140×55 mm, PP
Katalognr. 212-3911
Storage Boxes, PP, with Lid
Katalognr. 212-3911
Storage Boxes, PP, with Lid
Storage Boxes, PP, with Lid

with transparent hinged lid, 10×10 compartments, blu, 155×140×55 mm, PP

Preis
SFr. 11.50
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Top storage boxes made of polypropylene (PP) for dust-free storage of tubes.

  • Holds 100 tubes up to 12 mm diameter
  • Temperature resistant from −40 to +130 °C
Technische Daten
  • Colore:
    blu
  • Descrizione:
    with transparent hinged lid, 10×10 compartments
  • Materiale:
    PP
  • Seq:
    04
  • L×P×A:
    155×140×55 mm