Jars with overlapping lid
20 mm, 58 mm
Codice catalogo SEMA1711
Nome del fornitore: SEMADENI
Vai a
Product Details & Documents
Jars, made of PS, clear, with closing lid.
- Good chemical resistance
- Easy to clean
Suitable for ointments, powders, and granules.
Specifiche
- Ø est:20 mm
- Altezza:58 mm
- Seq:13