Jackets, Tyvek® 500, model PP33
Product Details & Documents
These jackets are made from Tyvek®, a flash spun high density PE fabric. Tyvek® is permeable to air and water vapour, but repels aerosols and water-based liquids. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
- Low-linting fabric
- Antistatic treatment (EN 1149-5) - on both sides
- With hood and zip closure
- Stitched seams (internal)
Typical applications include: pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up.
Colour: white
Certifications: (EU) 2016/425; partial body chemical protective clothing, cat III, Type PB [6-B]; EN 14126.
Packaging: 50 per box, bulk packed.
Specifiche
- Taglia dell'abbigliamento:XL
- Colore:bianco
- Seq:03
- Cat. No.:113-3886
- Genere:Unisex
- Modello:500 PP33
- Applicazioni per abiti:Chemical protection|Laboratory|Industrial
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



