Orders LinkContactUsLinkComponent
Screw caps for tubes
PP, 15-415
Codice catalogo DELT617200
Nome del fornitore:   DELTALAB
Screw caps for tubes
Codice catalogo DELT617200
Nome del fornitore:  DELTALAB
Screw caps for tubes
PP, 15-415
Price
SFr. 337.00
Log in per disponibilità
1 * 1.000 ST
Product Details & Documents
Screw caps made of polypropylene and aluminium with an internal liner made of rubber.
  • Both models of caps are fully autoclavable and are pharmaceutical quality
  • Excellent chemical resistance
  • Excellent temperature resistance
Screw caps for round bottom borosilicate glass tubes and flat bottom soda glass tubes.
Specifiche
  • Filettatura:
    15-415
  • Materiale:
    PP
  • Seq:
    02