Screw caps for tubes
PP, 15-415
Nome del fornitore: DELTALAB
Vai a
Product Details & Documents
Screw caps made of polypropylene and aluminium with an internal liner made of rubber.
- Both models of caps are fully autoclavable and are pharmaceutical quality
- Excellent chemical resistance
- Excellent temperature resistance
Screw caps for round bottom borosilicate glass tubes and flat bottom soda glass tubes.
Certificates
Cerca per numero di lottoSpecifiche
- Filettatura:15-415
- Materiale:PP
- Seq:02