Screw caps for tubes
PP, 13-415
Supplier: DELTALAB
Jump to:
Product Details & Documents
Screw caps made of polypropylene and aluminium with an internal liner made of rubber.
- Both models of caps are fully autoclavable and are pharmaceutical quality
- Excellent chemical resistance
- Excellent temperature resistance
Screw caps for round bottom borosilicate glass tubes and flat bottom soda glass tubes.
Documents
Certificates
Search by Lot NumberSpecifications
- Filettatura:13-415
- Materiale:PP
- Seq:02