Staining dishes, WHEATON®
Cover only, - mm
Codice catalogo 720-1033
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
Soda-lime glass and stainless steel.
- Handle permanently attached
- Hinged closure
- Easy insertion and removal of slides
The slotted rack hold 50 microscope slides 75×25 mm, 75×38 mm or 75×51 mm. Rack is made of non tarnishable, stainless steel that is resistant to staining solutions.
Specifiche
- Descrizione:Cover only
- L×P×A:- mm