Labels, hazard warning
Hazard labels: low temperature, PIC 323-TRI 050-B7541, Poliestere, nero on giallo, 50×43 mm, 7 labels/card
Katalognr. 116-0186
Anbieter: Brady
Direkt zu
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Zertifizierung: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Dokumente
Technische Daten
- Colore:nero on giallo
- Descrizione:Hazard labels: low temperature
- Dimensioni:50×43 mm
- Materiale:Poliestere
- Seq:17
- Modello:PIC 323-TRI 050-B7541
- Confezione:7 labels/card