Microplates, 96-well
Micropiastra da 96 pozzetti, mezzo volume, MICROLON® 200, 15 - 175 µl, 199 µl
Catalog # 738-0032
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
Crystal clear PS, without lid. Ideal for ELISA methodologies, these plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600) or medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200).
- Available with a choice of well profile: U-bottom, V-bottom, F-bottom (standard), F-bottom (chimney well), or half area
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Documents
Specifications
- Descrizione:Micropiastra da 96 pozzetti, mezzo volume, MICROLON® 200
- Seq:01
- Volume di lavoro:15 - 175 µl
- Volume totale:199 µl
- Cat. No.:738-0032
- Profilo del pozzetto:Mezzo volume
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:ISO 14001 (241002); ISO 50001 (241002)