Autoclave Bags
Autoclave resistant bags, 34 l, 750×500 mm, 30 µm
Supplier: DELTALAB
Jump to:
Product Details & Documents
Autoclave resistant bags, printed with the biohazard graphic symbol and instructions of use in five languages.
Caution: Do not close the bags hermetically when placing them in the autoclave at 121 °C.
Documents
Specifications
- Capacità:34 l
- Descrizione:Autoclave resistant bags
- L×L:750×500 mm
- Seq:03
- Spessore:30 µm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



