Petri dishes
Vented, 140 mm, 20 mm, Asettico, 12 bags of 10
Supplier: DELTALAB
Jump to:
Product Details & Documents
Vented petri dishes made from PS.
Specifications
- Ø:140 mm
- Confezione:12 bags of 10
- Sterilità:Asettico
- Versione:Vented
- Altezza:20 mm
- Seq:06