Adhesive mounting tapes
Glass silk tape, max. 450 °C, 12 mm
ISOP871904-000
: ISOPAD
Glass-silk adhesive tapes used for mounting.
- Larghezza:12 mm
- Descrizione:Glass silk tape, max. 450 °C
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



