Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Magnesio solfato ≥95%, TECHNICAL

Bidone in plastica, 25 kg
  25162.465
Magnesio solfato ≥95%, TECHNICAL
  25162.465
 :  7487-88-9
Magnesio solfato ≥95%, TECHNICAL
Magnesio solfato ≥95%, TECHNICAL

Bidone in plastica, 25 kg

 :  7487-88-9
 
666.00 SFr.
   
1 * 25 kg
 

 

  • Titolo:
    Min. 95 %

 

  • Tipo di conf.:
    Bidone in plastica
  • Conf:
    25 kg

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.