Container, EUROBOX
27 L, With one handle on lid, 400×300×340 mm
Catalog # ZARG40700
Supplier: ZARGES LEICHTBAU
Jump to:
Product Details & Documents
Aluminium, with lid.
- Lid with stainless steel hinges and holding straps
- Blue stacking corners ensure safe stacking
- Corner beads for additional dimensional stability
Eurobox has high resistance against corrosion due to stainless steel fittings. The peripheral foamed seal in the lid protects against dust and splash water.
Specifications
- Tipo:With one handle on lid
- Capacità:27 L
- Seq:01
- L×P×A:400×300×340 mm