Particle traps for use with PLOT columns
Particle trap for PoraPLOT columns, fused silica, Int.Ø×L: 0,53 mm × 2,5 m
Codice catalogo VARICP4017
Nome del fornitore: VARIAN
Vai a
Product Details & Documents
Though highly stabilised, it is impossible to guarantee that no particles will dislodge from the column wall. When used in valve-switching applications, the use of a particle trap can prevent scarring of the rotors.
Specifiche
- Descrizione:Particle trap for PoraPLOT columns, fused silica, Int.Ø×L: 0,53 mm × 2,5 m
- Seq:04