Instrument dishes
700 ml, 150×190×40 mm
Codice catalogo BURK4230-2070
Nome del fornitore: BURKLE
Vai a
Product Details & Documents
Trays for various uses. In laboratories, doctors’ surgeries, hospitals, pharmacies, the foodstuffs industry - wherever there is a need for clean storage of instruments, equipment, goods and precision parts.
- Melamine, white
- Tray without lid
Lid available as accessory.
Specifiche
- Capacità:700 ml
- L×P×A:150×190×40 mm