Capillary columns, VF-23ms
Capillare, VF-23ms, 0,50 µm, 0,53 mm, 30 m
Codice catalogo VARICP8831
Nome del fornitore: VARIAN
Vai a
Product Details & Documents
VF-23ms features a unique combination of high polarity and low bleed to enable more accurate analysis of very polar analytes.
- 100% bonded phase permits column rinsing to enhance column lifetime
- Fast run times improve productivity
- Operating temperature up to 260°C expands the application range
The enhanced stabilization of the VF-23ms permits splitless injection, column rinsing and temperatures up to 260 °C to be used.
Specifiche
- Formato della colonna:Capillare
- Spessore dello strato:0,50 µm
- Ø int.:0,53 mm
- Lunghezza:30 m
- Seq:05
- Fase:VF-23ms