Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Ferrules for gas chromatography

Ferrules for gas chromatography

Vespel ferrules, 1/8" to 0,425 mm short, for 0,25 mm ID columns
Codice catalogo VARICR213164
Nome del fornitore:   VARIAN
Ferrules for gas chromatography
Codice catalogo VARICR213164
Nome del fornitore:  VARIAN
Ferrules for gas chromatography
Ferrules for gas chromatography

Vespel ferrules, 1/8" to 0,425 mm short, for 0,25 mm ID columns

Price
Richiedi prezzo
Log in per disponibilità
1 * 10 ST
Product Details & Documents

Vespel/graphite and precision graphite ferrules for chromatography applications.

  • Does not require a back ferrule

Graphite ferrules are made from 100% high pure graphite with temperature limit of 450 °C. Ferrules are precision formed with a flash-free ID and may be reused if not over tightened. Graphite is ideal for glass-to-metal connections because it does not stick to glass. They used for various capillary columns.

Vespel/Graphite ferrules are made from 40% graphite and 60% high temperature DuPont polyimide, withstands 400 °C. They can be reused because they do not seize onto the column, ideal for GC/MS interface applications. Straight ferrules are used for the same dimension tubing.

Specifiche
  • Descrizione:
    Vespel ferrules, 1/8" to 0,425 mm short, for 0,25 mm ID columns
  • Seq:
    02