Capillary columns, CP-Wax™
on 7" cage, Capillare, CP-Wax 52 CB, 0,20 µm, 0,20 mm, 30 m
Codice catalogo VARICP7775
Nome del fornitore: VARIAN
Vai a
Product Details & Documents
The CP-Wax™ lines of chemically bonded columns offer a broad range of selectivities for method optimisation. The backbone of this series stationary phases is made of a polyethylene glycol.
Specifiche
- Formato della colonna:Capillare
- Spessore dello strato:0,20 µm
- Ø int.:0,20 mm
- Descrizione:on 7" cage
- Lunghezza:30 m
- Seq:4
- Fase:CP-Wax 52 CB