Particle traps for use with PLOT columns
Particle trap connectors for PoraPLOT columns, fused silica, 0,25/0,32 mm ID
Codice catalogo VARICP4788
Nome del fornitore: VARIAN
Vai a
Product Details & Documents
Though highly stabilised, it is impossible to guarantee that no particles will dislodge from the column wall. When used in valve-switching applications, the use of a particle trap can prevent scarring of the rotors.
Specifiche
- Descrizione:Particle trap connectors for PoraPLOT columns, fused silica, 0,25/0,32 mm ID
- Seq:06