Alclometasone Dipropionate, Reference Standards
Alclometasone Dipropionate, USP reference standard, 300 mg
Codice catalogo 1012757.
Nome del fornitore: USP
Alclometasone Dipropionate, Reference Standards
Codice catalogo 1012757.
Nome del fornitore: USP
Numero CAS:
Vai a
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
Specifiche
Specifiche
- Conf:300 mg
- Standard:USP reference standard
- Descrizione:Alclometasone Dipropionate
- Cat. No.:1012757.
- Formula:C₂₈H₃₇ClO₇
- Descrizione:Alclometasone Dipropionate
- CAS No.:66734-13-2



