Ammonio Methacrylate Copolymer , Reference Standards
Ammonio Methacrylate Copolymer Type B, USP reference standard, Vial, 500 mg
1029910.
: USP
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
- Tipo di conf.:Vial
- Conf:500 mg
- Standard:USP reference standard
- Descrizione:Ammonio Methacrylate Copolymer Type B
- Cat. No.:1029910.
- Formula:(C₉H₁₈NO₂.C₅H₈O₂.C₅H₈O₂.Cl)x
- Descrizione:Ammonio Methacrylate Copolymer
- CAS No.:33434-24-1



