Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Norgestrel, Reference Standards

Norgestrel, Reference Standards

Norgestrel, USP reference standard, Vial, 125 mg
Codice catalogo 1472000.
Nome del fornitore:   USP
Norgestrel, Reference Standards
Codice catalogo 1472000.
Nome del fornitore:  USP
Numero CAS:  
Norgestrel, Reference Standards

Norgestrel, USP reference standard, Vial, 125 mg

Numero CAS:  
Price
SFr. 383.00
Log in per disponibilità
1 * 125 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifiche

Specifiche

  • Tipo di conf.:
    Vial
  • Conf:
    125 mg
  • Standard:
    USP reference standard
  • Descrizione:
    Norgestrel
  • Cat. No.:
    1472000.
  • Formula:
    C₂₁H₂₈O₂
  • Descrizione:
    Norgestrel
  • CAS No.:
    6533-00-2

Specifiche