Nevirapine, Reference Standards
Nevirapine Anhydrous, USP reference standard, Vial, 100 mg
Codice catalogo 1460703.
Nome del fornitore: USP
Vai a
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifiche
Specifiche
- Tipo di conf.:Vial
- Conf:100 mg
- Standard:USP reference standard
- Descrizione:Nevirapine Anhydrous
- Cat. No.:1460703.
- Formula:C₁₅H₁₄N₄O
- Descrizione:Nevirapine Anhydrous
- CAS No.:129618-40-2



